Gene Transcripts

Gene Symbol
LCE6A
Entrez Gene ID
448835
Gene Name
late cornified envelope 6A
Gene Aliases
C1orf44
Chromosome Location
Chr. 1: 152842830 - 152844211 on Build GRCh38
UniGene ID
Hs.62927
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128600.1 NP_001122072.1 2 629
XM_017001327.1 XP_016856816.1 2 550
GenBank mRNA BG210316.1 - 1 476
DQ991251.1 - 2 629

View Full Product Details