Gene Transcripts

Gene Symbol
SPANXN4
Entrez Gene ID
441525
Gene Name
SPANX family member N4
Gene Aliases
CT11.9
Chromosome Location
Chr. X: 143025918 - 143038637 on Build GRCh38
UniGene ID
Hs.535082
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009613.2 NP_001009613.1 2 256
XM_017029543.1 XP_016885032.1 2 246
XM_017029544.1 XP_016885033.1 2 243
GenBank mRNA BC130503.1 - 2 256
BC130505.1 - 2 256 40146536
DQ336126.1 - 2 159
DQ336127.1 - 2 159

View Full Product Details