Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001205049.1 | NP_001191978.1 | 3 | 363 | |
NM_001277245.1 | NP_001264174.1 | 3 | 426 | ||
NM_001277246.1 | NP_001264175.1 | 3 | 359 | ||
NM_001277247.1 | NP_001264176.1 | 3 | 237 | ||
NM_013455.3 | NP_038483.1 | 3 | 363 | ||
GenBank mRNA | AK006695.1 | - | 3 | 363 | |
AK160427.1 | - | 3 | 310 | ||
BC103577.1 | - | 3 | 326 | ||
BC103578.1 | - | 3 | 326 | ||
BC103579.1 | - | 3 | 389 | 40045630 | |
BC103580.1 | - | 3 | 326 | 40045632 | |
D00754.1 | - | 3 | 323 | ||
M85170.1 | - | 2 | 326 | ||
X52466.1 | - | 2 | 326 |