Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008743.2 | NP_032769.2 | 3 | 540 | |
GenBank mRNA | AB006812.1 | - | 3 | 486 | |
AK033701.1 | - | 3 | 1548 | ||
AV377313.2 | - | 1 | 128 | ||
BC138852.1 | - | 3 | 503 | 8861874 | |
BC138855.1 | - | 3 | 503 | 8861877 | |
BC145443.1 | - | 3 | 412 | ||
CX565655.1 | - | 3 | 519 | ||
Y09688.1 | - | 3 | 512 |