Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001205322.1 | NP_001192251.1 | 3 | 351 | |
NM_008864.3 | NP_032890.2 | 3 | 348 | ||
GenBank mRNA | AF525162.1 | - | 322 | ||
AK082929.1 | - | 3 | 347 | ||
BC064736.1 | - | 3 | 331 | 30437709 | |
M35662.1 | - | 3 | 328 |