Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011171.2 | NP_035301.2 | 3, 4 | 787 | |
GenBank mRNA | AK136541.1 | - | 3, 4 | 648 | |
AK143719.1 | - | 3, 4 | 889 | ||
AK167302.1 | - | 3, 4 | 727 | ||
CT010175.1 | - | 3, 4 | 606 | ||
L39017.1 | - | 3, 4 | 661 | ||
X63748.2 | - | 3, 4 | 693 |