Gene Transcripts

Gene Symbol
Vmn2r88
Entrez Gene ID
669149
Gene Name
vomeronasal 2, receptor 88
Gene Aliases
V2r13, V2r3
Chromosome Location
Chr. 14: 51411001 - 51419527 on Build GRCm38
UniGene ID
Mm.377117
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011686.1 NP_035816.1 2 351
GenBank mRNA AF011413.1 - 2 213
AF011423.1 - 2 278

View Full Product Details