Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289480.1 | NP_001276409.1 | 9 | 1308 | |
NM_013709.5 | NP_038737.1 | 10 | 1422 | ||
GenBank mRNA | AK141199.1 | - | 10 | 1423 | |
AK168483.1 | - | 11 | 1048 | ||
BC010296.1 | - | 9 | 870 | 3154966 | |
BC058557.1 | - | 9 | 1285 | 6313231 | |
BC061488.1 | - | 9 | 2022 | 4483634 | |
BC110043.1 | - | 10 | 1400 | 5149633 | |
D85926.1 | - | 10 | 905 |