Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021419.1 | NP_067394.1 | 2 | 285 | |
GenBank mRNA | AJ242721.1 | - | 229 | ||
AK076180.1 | - | 2 | 265 | ||
AK147168.1 | - | 2 | 307 | ||
AK219887.1 | - | 2 | 60 | ||
BC021778.1 | - | 2 | 285 | 4239442 | |
CT010295.1 | - | 2 | 148 |