Gene Transcripts

Gene Symbol
Rps6ka6
Entrez Gene ID
67071
Gene Name
ribosomal protein S6 kinase polypeptide 6
Gene Aliases
2610524K04Rik, RSK4, S6K-alpha-6
Chromosome Location
Chr. X: 111387837 - 111538097 on Build GRCm38
UniGene ID
Mm.37617
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025949.3 NP_080225.2 17 1688
GenBank mRNA AK012150.2 - 6 440
AK049349.1 - 15 1439
BC054113.1 - 15 1361 30015425

View Full Product Details