Gene Transcripts

Gene Symbol
DNAJB9
Entrez Gene ID
4189
Gene Name
DnaJ heat shock protein family (Hsp40) member B9
Gene Aliases
ERdj4, MDG-1, MDG1, MST049, MSTP049
Chromosome Location
Chr. 7: 108569745 - 108574850 on Build GRCh38
UniGene ID
Hs.732494
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012328.2 NP_036460.1 3 600
GenBank mRNA AB026908.1 - 3 433
AF083247.1 - 3 434
AF093824.1 - 3 400
AK092204.1 - 4 458
AL080081.1 - 3 480
AY359045.1 - 2 254
BC028912.1 - 3 414 4943933
CR533475.1 - 2 231

View Full Product Details