Gene Transcripts

Gene Symbol
Agmat
Entrez Gene ID
75986
Gene Name
agmatine ureohydrolase (agmatinase)
Gene Aliases
5033405N08Rik
Chromosome Location
Chr. 4: 141746675 - 141759263 on Build GRCm38
UniGene ID
Mm.30032
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081408.1 NP_001074877.1 2 571
GenBank mRNA AK017155.1 - 2 470

View Full Product Details