Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031392.2 | NP_113569.1 | 2 | 873 | |
GenBank mRNA | AB041854.1 | - | 2 | 942 | |
AF348591.1 | - | 2 | 869 | ||
AK037181.1 | - | 2 | 850 | ||
AK046119.1 | - | 2 | 840 | ||
AK052235.1 | - | 2 | 814 | ||
AK146617.1 | - | 2 | 830 | ||
AK163752.1 | - | 2 | 806 | ||
AK165935.1 | - | 2 | 837 | ||
AK169357.1 | - | 2 | 874 | ||
BC050894.1 | - | 2 | 873 | 5708880 | |
BC054367.1 | - | 2 | 801 | 5346943 |