Gene Transcripts

Gene Symbol
Mbd6
Entrez Gene ID
110962
Gene Name
methyl-CpG binding domain protein 6
Gene Aliases
D10Wsu93e
Chromosome Location
Chr. 10: 127281956 - 127290719 on Build GRCm38
UniGene ID
Mm.372826
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033072.2 NP_149063.2 7 2178
GenBank mRNA AK158797.1 - 7 2302
BC059015.1 - 8 1890
BC096027.1 - 4 1920

View Full Product Details