Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193511.1 | NP_001180440.1 | 7 | 1506 | |
NM_006301.3 | NP_006292.3 | 8 | 1407 | ||
XM_005269138.3 | XP_005269195.1 | 7 | 1434 | ||
XM_005269140.2 | XP_005269197.1 | 7 | 1347 | ||
XM_006719588.3 | XP_006719651.1 | 7 | 1328 | ||
XM_011538725.2 | XP_011537027.1 | 7 | 2311 | ||
XM_017019956.1 | XP_016875445.1 | 8 | 1335 | ||
GenBank mRNA | AB209453.1 | - | 2 | 2357 | |
AK094195.1 | - | 7 | 1302 | ||
AK314530.1 | - | 7 | 1654 | ||
BC050050.1 | - | 7 | 1507 | 4824512 | |
U07358.1 | - | 8 | 1234 |