Gene Transcripts
- Gene Symbol
- TBC1D26
- Entrez Gene ID
- 353149
- Gene Name
- TBC1 domain family member 26
- Gene Aliases
- Chromosome Location
- Chr. 17: 15732277 - 15744784 on Build GRCh38
- UniGene ID
- Hs.434123
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC028414.1 |
- |
13 |
3805 |
|
BC047400.1 |
- |
16 |
1561 |
5297645 |
View Full Product Details