Gene Transcripts
- Gene Symbol
- PPFIA2
- Entrez Gene ID
- 8499
- Gene Name
- PTPRF interacting protein alpha 2
- Gene Aliases
- Chromosome Location
- Chr. 12: 81257975 - 81759748 on Build GRCh38
- UniGene ID
- Hs.506216
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001220473.2 |
NP_001207402.1
|
6, 7 |
948 |
|
NM_001220474.2 |
NP_001207403.1
|
5, 6 |
894 |
|
NM_001220475.2 |
NP_001207404.1
|
6, 7 |
760 |
|
NM_001220476.2 |
NP_001207405.1
|
6, 7 |
760 |
|
NM_001220477.2 |
NP_001207406.1
|
4, 5 |
767 |
|
NM_001282536.1 |
NP_001269465.1
|
4, 5 |
455 |
|
NM_003625.4 |
NP_003616.2
|
7, 8 |
811 |
|
XM_006719670.3 |
XP_006719733.1
|
4, 5 |
8003 |
|
XM_011538906.2 |
XP_011537208.1
|
5, 6 |
888 |
|
XM_011538907.2 |
XP_011537209.1
|
5, 6 |
888 |
|
XM_017020079.1 |
XP_016875568.1
|
7, 8 |
795 |
|
XM_017020080.1 |
XP_016875569.1
|
6, 7 |
942 |
|
XM_017020081.1 |
XP_016875570.1
|
6, 7 |
942 |
|
XM_017020082.1 |
XP_016875571.1
|
7, 8 |
795 |
|
XM_017020083.1 |
XP_016875572.1
|
6, 7 |
942 |
|
XM_017020084.1 |
XP_016875573.1
|
6, 7 |
942 |
|
XM_017020085.1 |
XP_016875574.1
|
6, 7 |
942 |
|
XM_017020086.1 |
XP_016875575.1
|
7, 8 |
795 |
|
XM_017020087.1 |
XP_016875576.1
|
6, 7 |
929 |
|
XM_017020088.1 |
XP_016875577.1
|
6, 7 |
1175 |
|
XM_017020089.1 |
XP_016875578.1
|
5, 6 |
888 |
|
XM_017020090.1 |
XP_016875579.1
|
7, 8 |
795 |
|
XM_017020091.1 |
XP_016875580.1
|
6, 7 |
942 |
|
XM_017020092.1 |
XP_016875581.1
|
6, 7 |
942 |
|
XM_017020093.1 |
XP_016875582.1
|
6, 7 |
942 |
|
XM_017020095.1 |
XP_016875584.1
|
6, 7 |
942 |
|
XM_017020097.1 |
XP_016875586.1
|
7, 8 |
795 |
|
XM_017020098.1 |
XP_016875587.1
|
7, 8 |
795 |
|
XM_017020099.1 |
XP_016875588.1
|
5, 6 |
888 |
|
XM_017020100.1 |
XP_016875589.1
|
6, 7 |
942 |
|
XM_017020101.1 |
XP_016875590.1
|
7, 8 |
795 |
|
XM_017020103.1 |
XP_016875592.1
|
6, 7 |
942 |
|
XM_017020104.1 |
XP_016875593.1
|
6, 7 |
942 |
|
XM_017020106.1 |
XP_016875595.1
|
6, 7 |
942 |
|
XM_017020107.1 |
XP_016875596.1
|
7, 8 |
795 |
|
XM_017020108.1 |
XP_016875597.1
|
7, 8 |
795 |
|
XM_017020109.1 |
XP_016875598.1
|
6, 7 |
942 |
|
XM_017020111.1 |
XP_016875600.1
|
6, 7 |
942 |
|
XM_017020112.1 |
XP_016875601.1
|
4, 5 |
767 |
|
XM_017020113.1 |
XP_016875602.1
|
4, 5 |
767 |
|
XM_017020114.1 |
XP_016875603.1
|
4, 5 |
767 |
|
XM_017020115.1 |
XP_016875604.1
|
4, 5 |
767 |
|
XM_017020116.1 |
XP_016875605.1
|
4, 5 |
767 |
|
XM_017020118.1 |
XP_016875607.1
|
5, 6 |
742 |
|
XM_017020119.1 |
XP_016875608.1
|
4, 5 |
767 |
|
XM_017020120.1 |
XP_016875609.1
|
4, 5 |
455 |
|
GenBank mRNA |
AB210009.1 |
- |
6, 7 |
946 |
|
AF034799.1 |
- |
7, 8 |
819 |
|
AK126971.1 |
- |
5, 6 |
854 |
|
AK297372.1 |
- |
5, 6 |
742 |
|
AK299853.1 |
- |
4, 5 |
716 |
|
AK302658.1 |
- |
4, 5 |
426 |
|
AK307524.1 |
- |
7, 8 |
795 |
|
BC104912.1 |
- |
6, 7 |
744 |
8143915 |
BC143485.1 |
- |
6, 7 |
744 |
9051993 |
DB083201.1 |
- |
4, 5 |
455 |
|
View Full Product Details