Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005415.4 | NP_005406.3 | 8 | 1744 | |
XM_017004768.1 | XP_016860257.1 | 8 | 1493 | ||
XM_017004769.1 | XP_016860258.1 | 9 | 3971 | ||
XM_017004770.1 | XP_016860259.1 | 9 | 1774 | ||
XM_017004771.1 | XP_016860260.1 | 10 | 1810 | ||
GenBank mRNA | AK123420.1 | - | 3 | 1282 | |
AK312738.1 | - | 8 | 1663 | ||
BC019944.1 | - | 8 | 1651 | 3918690 | |
BC075818.1 | - | 2 | 253 | ||
BX647555.1 | - | 1 | 997 | ||
L20859.1 | - | 8 | 1575 |