Gene Transcripts

Gene Symbol
NPTXR
Entrez Gene ID
23467
Gene Name
neuronal pentraxin receptor
Gene Aliases
NPR
Chromosome Location
Chr. 22: 38818451 - 38844012 on Build GRCh38
UniGene ID
Hs.91622
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014293.3 NP_055108.2 5 1473
GenBank mRNA AL161974.1 - 5 787
AL162057.1 - 1 114

View Full Product Details