Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014564.4 | NP_055379.1 | 6 | 2282 | |
NM_178138.5 | NP_835258.1 | 6 | 2250 | ||
XM_005263410.1 | XP_005263467.1 | 6 | 2564 | ||
XM_017015168.1 | XP_016870657.1 | 5 | 2061 | ||
GenBank mRNA | AB593062.1 | - | 6 | 2250 | |
AB593063.1 | - | 6 | 2249 | ||
AB593064.1 | - | 6 | 2564 | ||
AB593065.1 | - | 5 | 2957 | ||
AB593066.1 | - | 6 | 2282 | ||
AB593067.1 | - | 6 | 2247 | ||
AF156888.1 | - | 7 | 2045 | ||
AF156889.1 | - | 6 | 2267 |