Gene Transcripts

Gene Symbol
LRRC39
Entrez Gene ID
127495
Gene Name
leucine rich repeat containing 39
Gene Aliases
Chromosome Location
Chr. 1: 100148448 - 100178273 on Build GRCh38
UniGene ID
Hs.44277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256385.1 NP_001243314.1 5, 6 652
NM_001256386.1 NP_001243315.1 5, 6 652
NM_001256387.1 NP_001243316.1 4, 5 595
NM_144620.3 NP_653221.1 5, 6 635
GenBank mRNA AK096892.1 - 5, 6 578
AL833417.1 - 5, 6 600
AY358131.1 - 4, 5 568
BC009613.1 - 5, 6 577 4245944

View Full Product Details