Gene Transcripts

Gene Symbol
NKX6-3
Entrez Gene ID
157848
Gene Name
NK6 homeobox 3
Gene Aliases
NKX6.3
Chromosome Location
Chr. 8: 41645177 - 41650818 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152568.3 NP_689781.1 2 1588
XM_017013143.1 XP_016868632.1 3 2300
GenBank mRNA AK057898.1 - 3 1545

View Full Product Details