Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144026.2 | NP_001137498.1 | 3 | 343 | |
NM_001144027.2 | NP_001137499.1 | 3 | 343 | ||
NM_001144028.2 | NP_001137500.1 | 3 | 343 | ||
NM_014434.3 | NP_055249.1 | 3 | 343 | ||
XM_011518545.1 | XP_011516847.1 | 3 | 333 | ||
XM_011518547.1 | XP_011516849.1 | 3 | 133 | ||
XM_011518548.2 | XP_011516850.1 | 3 | 322 | ||
XM_017014626.1 | XP_016870115.1 | 3 | 343 | ||
XM_017014627.1 | XP_016870116.1 | 3 | 343 | ||
GenBank mRNA | AF199509.1 | - | 3 | 279 | |
AK026089.1 | - | 3 | 315 | ||
AK074403.1 | - | 3 | 304 | ||
AK225995.1 | - | 3 | 314 | ||
AK290026.1 | - | 3 | 340 | ||
AY077845.1 | - | 3 | 261 | ||
BC015735.1 | - | 3 | 279 | 4869561 | |
BC078668.1 | - | 3 | 279 | ||
BC093782.1 | - | 3 | 260 | 7939627 | |
BC111943.1 | - | 3 | 260 | 8327411 | |
DA408370.1 | - | 3 | 343 | ||
DA858250.1 | - | 3 | 343 |