Gene Transcripts
- Gene Symbol
- ST6GALNAC1
- Entrez Gene ID
- 55808
- Gene Name
- ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1
- Gene Aliases
- HSY11339, SIAT7A, ST6GalNAcI, STYI
- Chromosome Location
- Chr. 17: 76617769 - 76643838 on Build GRCh38
- UniGene ID
- Hs.105352
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018414.4 |
NP_060884.1
|
1, 2 |
339 |
|
NR_110309.1 |
|
1, 2 |
339 |
|
XM_011524995.2 |
XP_011523297.1
|
1, 2 |
259 |
|
XM_011524996.2 |
XP_011523298.1
|
1, 2 |
259 |
|
XM_017024842.1 |
XP_016880331.1
|
1, 2 |
259 |
|
XM_017024843.1 |
XP_016880332.1
|
1, 2 |
259 |
|
GenBank mRNA |
AK310906.1 |
- |
1, 2 |
275 |
|
AY096001.1 |
- |
1, 2 |
139 |
|
AY358918.1 |
- |
1, 2 |
284 |
|
BC022462.1 |
- |
1, 2 |
194 |
4816713 |
DA594489.1 |
- |
1, 2 |
339 |
|
Y11339.2 |
- |
1, 2 |
199 |
|
View Full Product Details