Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001017403.1 | NP_001017403.1 | 3 | 333 | |
NM_001017404.1 | NP_001017404.1 | 3 | 204 | ||
NM_021636.2 | NP_067649.2 | 3 | 220 | ||
XM_005245404.3 | XP_005245461.1 | 3 | 277 | ||
XM_011509838.2 | XP_011508140.1 | 3 | 278 | ||
XM_011509839.2 | XP_011508141.1 | 3 | 279 | ||
XM_011509840.2 | XP_011508142.1 | 3 | 278 | ||
XM_017001996.1 | XP_016857485.1 | 3 | 279 | ||
GenBank mRNA | AB049405.1 | - | 3 | 218 | |
AF190501.1 | - | 3 | 204 | ||
AY358119.1 | - | 3 | 261 | ||
BC047905.1 | - | 3 | 203 | 5212508 |