Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001319051.1 | NP_001305980.1 | 5 | 2006 | |
NM_054110.4 | NP_473451.3 | 5 | 1651 | ||
XM_005264852.4 | XP_005264909.1 | 5 | 1183 | ||
XM_017005682.1 | XP_016861171.1 | 5 | 9064 | ||
XM_017005683.1 | XP_016861172.1 | 5 | 1214 | ||
XM_017005684.1 | XP_016861173.1 | 5 | 1214 | ||
GenBank mRNA | AB078149.1 | - | 5 | 1176 | |
AK094780.1 | - | 8 | 1224 | ||
AK312425.1 | - | 5 | 1652 | ||
AL831925.1 | - | 5 | 1679 | ||
AL832575.2 | - | 5 | 1153 | ||
AL832736.1 | - | 5 | 1621 | ||
AY035399.1 | - | 5 | 1459 | ||
AY358443.1 | - | 5 | 1637 | ||
BC014789.1 | - | 5 | 1601 | 4212702 |