Gene Transcripts

Gene Symbol
ST6GALNAC3
Entrez Gene ID
256435
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 3
Gene Aliases
PRO7177, SIAT7C, ST6GALNACIII, STY
Chromosome Location
Chr. 1: 76074684 - 76632880 on Build GRCh38
UniGene ID
Hs.337040
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160011.1 NP_001153483.1 3 606
NM_152996.2 NP_694541.2 3 606
XM_006710553.3 XP_006710616.1 4 602
XM_017000937.1 XP_016856426.1 3 3748
XM_017000938.1 XP_016856427.1 3 3760
XM_017000939.1 XP_016856428.1 3 3758
XM_017000940.1 XP_016856429.1 4 865
XM_017000941.1 XP_016856430.1 3 1520
XM_017000942.1 XP_016856431.1 3 1325
GenBank mRNA AJ507291.1 - 3 464
AK091215.1 - 3 591
AK297363.1 - 2 375
AY358540.1 - 3 565
BC039520.1 - 2 317
BC059363.1 - 3 577 30341233

View Full Product Details