Gene Transcripts
- Gene Symbol
- PKD2L2
- Entrez Gene ID
- 27039
- Gene Name
- polycystin 2 like 2, transient receptor potential cation channel
- Gene Aliases
- TRPP5
- Chromosome Location
- Chr. 5: 137889420 - 137942747 on Build GRCh38
- UniGene ID
- Hs.716884
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001258448.1 |
NP_001245377.1
|
12, 13 |
1663 |
|
NM_001258449.1 |
NP_001245378.1
|
10, 11 |
1426 |
|
NM_001300921.1 |
NP_001287850.1
|
12, 13 |
1729 |
|
NM_014386.3 |
NP_055201.2
|
12, 13 |
1729 |
|
XM_011543318.2 |
XP_011541620.1
|
11, 12 |
1643 |
|
XM_011543321.1 |
XP_011541623.1
|
12, 13 |
1663 |
|
XM_017009344.1 |
XP_016864833.1
|
11, 12 |
1853 |
|
XM_017009345.1 |
XP_016864834.1
|
11, 12 |
1523 |
|
GenBank mRNA |
AF118125.1 |
- |
12, 13 |
1696 |
|
AF182034.1 |
- |
12, 13 |
1699 |
|
AK301924.1 |
- |
12, 13 |
1632 |
|
BC044581.1 |
- |
10, 11 |
1425 |
4830805 |
View Full Product Details