Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020318.2 | NP_064714.2 | 2 | 1260 | |
NM_021936.2 | NP_068755.2 | 2 | 1260 | ||
XM_005245422.3 | XP_005245479.1 | 2 | 1307 | ||
XM_011509857.1 | XP_011508159.1 | 3 | 1177 | ||
XM_011509858.2 | XP_011508160.1 | 2 | 1520 | ||
XM_017002023.1 | XP_016857512.1 | 3 | 1358 | ||
XM_017002024.1 | XP_016857513.1 | 3 | 1405 | ||
XM_017002025.1 | XP_016857514.1 | 2 | 1511 | ||
GenBank mRNA | AF311940.1 | - | 1 | 96 | |
AF342989.1 | - | 2 | 1260 | ||
AF342990.1 | - | 2 | 1260 | ||
AJ278348.1 | - | 1 | 59 | ||
BC117193.1 | - | 1 | 868 | ||
BC117195.1 | - | 1 | 869 | ||
BC152552.1 | - | 2 | 1248 |