Gene Transcripts

Gene Symbol
RBL1
Entrez Gene ID
5933
Gene Name
RB transcriptional corepressor like 1
Gene Aliases
CP107, PRB1, p107
Chromosome Location
Chr. 20: 36996349 - 37096215 on Build GRCh38
UniGene ID
Hs.207745
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323281.1 NP_001310210.1 2 504
NM_001323282.1 NP_001310211.1 2 504
NM_002895.4 NP_002886.2 2 504
NM_183404.3 NP_899662.1 2 504
XM_006723842.2 XP_006723905.1 2 280
XM_011528955.2 XP_011527257.1 2 280
XM_011528958.2 XP_011527260.1 2 280
XM_017027992.1 XP_016883481.1 2 318
GenBank mRNA AK290380.1 - 2 280
AK308096.1 - 2 264
AK310870.1 - 2 289
BC032247.1 - 2 287 4521266
DC375054.1 - 2 506
L14812.1 - 2 285

View Full Product Details