Gene Transcripts

Gene Symbol
ZFP41
Entrez Gene ID
286128
Gene Name
ZFP41 zinc finger protein
Gene Aliases
ZNF753, zfp-41
Chromosome Location
Chr. 8: 143246821 - 143262705 on Build GRCh38
UniGene ID
Hs.668016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271156.2 NP_001258085.2 2 1382
NM_173832.5 NP_776193.2 2 668
GenBank mRNA AK096024.1 - 2 657
AL831951.1 - 2 1093
BC034608.1 - 2 1382 4827717
CR992458.1 - 1 114
DA216172.1 - 1 312

View Full Product Details