Gene Transcripts

Gene Symbol
BCL6B
Entrez Gene ID
255877
Gene Name
B-cell CLL/lymphoma 6B
Gene Aliases
BAZF, ZBTB28, ZNF62
Chromosome Location
Chr. 17: 7023050 - 7029642 on Build GRCh38
UniGene ID
Hs.22575
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181844.3 NP_862827.1 9 2277
GenBank mRNA AK024695.1 - 1 440
AK122893.1 - 9 2271
BC059404.1 - 9 2276 30342006

View Full Product Details