Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005630.2 | NP_005621.2 | 8 | 1244 | |
XM_017007075.1 | XP_016862564.1 | 8 | 1187 | ||
XM_017007076.1 | XP_016862565.1 | 8 | 1187 | ||
XM_017007077.1 | XP_016862566.1 | 9 | 1232 | ||
GenBank mRNA | AK303550.1 | - | 4 | 550 | |
BC041140.1 | - | 8 | 1073 | 5751516 | |
BC051347.1 | - | 8 | 1153 | 6481104 | |
U70867.1 | - | 8 | 1053 |