Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001127218.2 | NP_001120690.1 | 4 | 657 | |
NM_001256879.1 | NP_001243808.1 | 5 | 1018 | ||
NM_006230.3 | NP_006221.2 | 4 | 484 | ||
XM_011515431.1 | XP_011513733.1 | 4 | 480 | ||
GenBank mRNA | AK292347.1 | - | 5 | 1018 | |
AK298168.1 | - | 4 | 348 | ||
AK312294.1 | - | 4 | 617 | ||
AU117477.2 | - | 4 | 462 | ||
BC000459.2 | - | 4 | 447 | 2822169 | |
BC030001.2 | - | 3 | 424 | ||
BP224472.1 | - | 4 | 485 | ||
U21090.1 | - | 4 | 446 |