Gene Transcripts

Gene Symbol
OSBPL1A
Entrez Gene ID
114876
Gene Name
oxysterol binding protein like 1A
Gene Aliases
ORP-1, ORP1, OSBPL1B
Chromosome Location
Chr. 18: 24162045 - 24397882 on Build GRCh38
UniGene ID
Hs.370725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242508.1 NP_001229437.1 16 1733
NM_018030.4 NP_060500.3 14 2102
NM_080597.3 NP_542164.2 28 2989
XM_006722380.3 XP_006722443.1 27 2902
XM_006722382.3 XP_006722445.1 27 2867
XM_017025530.1 XP_016881019.1 28 3281
XM_017025531.1 XP_016881020.1 26 2591
XM_017025533.1 XP_016881022.1 14 1585
XM_017025534.1 XP_016881023.1 14 4113
GenBank mRNA AF274714.1 - 14 2102
AF323726.2 - 28 2877
AF392449.1 - 14 1723
AF392450.1 - 28 2946
AK096358.1 - 28 2989
AK292895.1 - 29 2921
AK301862.1 - 16 1733
AK314552.1 - 14 1553
BC007004.1 - 2 101
BC063420.1 - 14 1788
BC073854.1 - 12 1309
BC098384.1 - 14 1781
BC136452.1 - 28 2886 9020441
BC144117.1 - 28 2886 9052637

View Full Product Details