Gene Transcripts

Gene Symbol
NLRX1
Entrez Gene ID
79671
Gene Name
NLR family member X1
Gene Aliases
CLR11.3, DLNB26, NOD26, NOD5, NOD9
Chromosome Location
Chr. 11: 119168334 - 119184016 on Build GRCh38
UniGene ID
Hs.524082
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282143.1 NP_001269072.1 5 1060
NM_001282144.1 NP_001269073.1 5 1468
NM_001282358.1 NP_001269287.1 5 992
NM_024618.3 NP_078894.2 5 1087
XM_005271669.1 XP_005271726.1 5 1311
XM_005271670.1 XP_005271727.1 4 964
XM_005271671.1 XP_005271728.1 4 1138
XM_005271672.1 XP_005271729.1 4 961
XM_006718904.1 XP_006718967.1 5 1345
XM_011542980.1 XP_011541282.1 5 975
XM_017018294.1 XP_016873783.1 5 1345
GenBank mRNA AB094095.1 - 5 1071
AK056454.1 - 4 785
AK095247.1 - 3 272
AK291716.1 - 5 1054
AK303002.1 - 4 961
AY245437.1 - 4 830
BC013199.1 - 1 191
BC110890.1 - 5 1433 4937963
BX647705.1 - 5 1045
EF452237.1 - 4 830

View Full Product Details