Gene Transcripts

Gene Symbol
FAM208A
Entrez Gene ID
23272
Gene Name
family with sequence similarity 208 member A
Gene Aliases
C3orf63, RAP140, se89-1
Chromosome Location
Chr. 3: 56620132 - 56683242 on Build GRCh38
UniGene ID
Hs.116877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001112736.1 NP_001106207.1 13 1604
NM_015224.3 NP_056039.2 6 955
XM_005264999.1 XP_005265056.1 13 1739
XM_006713077.2 XP_006713140.1 13 1739
XM_006713078.2 XP_006713141.1 7 593
XM_011533552.2 XP_011531854.1 13 1739
XM_011533553.2 XP_011531855.1 12 1160
XM_017006030.1 XP_016861519.1 13 1514
XM_017006031.1 XP_016861520.1 13 1739
XM_017006032.1 XP_016861521.1 12 1160
GenBank mRNA AF180425.2 - 6 1239
AK094486.1 - 13 1734
AK307599.1 - 5 417
BC029354.1 - 6 955
BC070096.1 - 13 1602
BC129987.1 - 5 380 40129237
BX648955.1 - 12 1086

View Full Product Details