Gene Transcripts

Gene Symbol
MREG
Entrez Gene ID
55686
Gene Name
melanoregulin
Gene Aliases
DSU, WDT2
Chromosome Location
Chr. 2: 215939097 - 216013623 on Build GRCh38
UniGene ID
Hs.620391
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018000.2 NP_060470.2 4 728
XM_011511468.2 XP_011509770.1 5 728
GenBank mRNA AF068290.1 - 3 227
AK000978.1 - 4 698
BC032747.1 - 4 737 5573149
BC082990.1 - 4 718 6159624
CX781801.1 - 4 723

View Full Product Details