Gene Transcripts

Gene Symbol
LRRC8E
Entrez Gene ID
80131
Gene Name
leucine rich repeat containing 8 family member E
Gene Aliases
Chromosome Location
Chr. 19: 7888505 - 7902023 on Build GRCh38
UniGene ID
Hs.501511
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001268284.2 NP_001255213.1 4 561
NM_001268285.2 NP_001255214.1 2 275
NM_025061.5 NP_079337.2 3 418
XM_011528319.2 XP_011526621.1 3 1146
GenBank mRNA AK091134.1 - 3 418
BC070089.1 - 3 418 30346745
BX538180.1 - 3 408
CD109368.1 - 3 445
CF593782.1 - 4 544
DA447942.1 - 2 240

View Full Product Details