Gene Transcripts

Gene Symbol
NINL
Entrez Gene ID
22981
Gene Name
ninein like
Gene Aliases
NLP
Chromosome Location
Chr. 20: 25452697 - 25585531 on Build GRCh38
UniGene ID
Hs.631508
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318226.1 NP_001305155.1 15, 16 2013
NM_025176.5 NP_079452.3 15, 16 2013
XM_011529186.2 XP_011527488.1 15, 16 2651
XM_011529187.2 XP_011527489.1 15, 16 2636
XM_011529188.1 XP_011527490.1 15, 16 2028
XM_011529189.1 XP_011527491.1 15, 16 2036
XM_011529190.1 XP_011527492.1 14, 15 1951
XM_011529191.1 XP_011527493.1 11, 12 1543
XM_011529192.1 XP_011527494.1 8, 9 2463
XM_011529193.1 XP_011527495.1 14, 15 1951
XM_011529194.2 XP_011527496.1 6, 7 1732
XM_011529195.2 XP_011527497.1 5, 6 557
XM_011529197.2 XP_011527499.1 7, 8 926
XM_017027732.1 XP_016883221.1 16, 17 3970
XM_017027733.1 XP_016883222.1 16, 17 2082
XM_017027734.1 XP_016883223.1 9, 10 1117
XM_017027735.1 XP_016883224.1 5, 6 562
GenBank mRNA AB023197.1 - 15, 16 1999
AK093569.1 - 5, 6 1396
AK309298.1 - 15, 16 1962
BC036380.2 - 14, 15 1951 5268454
EU718622.1 - 15, 16 1999

View Full Product Details