Gene Transcripts

Gene Symbol
ZGRF1
Entrez Gene ID
55345
Gene Name
zinc finger GRF-type containing 1
Gene Aliases
C4orf21
Chromosome Location
Chr. 4: 112539333 - 112636995 on Build GRCh38
UniGene ID
Hs.380346
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018392.4 NP_060862.3 5 451
XM_005263115.3 XP_005263172.1 5 506
XM_011532091.2 XP_011530393.1 5 451
XM_011532092.2 XP_011530394.1 4 331
XM_011532093.2 XP_011530395.1 5 451
XM_011532094.2 XP_011530396.1 5 451
XM_011532096.2 XP_011530398.1 5 451
XM_017008369.1 XP_016863858.1 4 446
XM_017008370.1 XP_016863859.1 5 451
XM_017008371.1 XP_016863860.1 4 331
XM_017008372.1 XP_016863861.1 5 360
XM_017008373.1 XP_016863862.1 4 297
GenBank mRNA AK002193.1 - 5 368
AK299831.1 - 5 451
AK302613.1 - 6 676
AL708767.1 - 5 374
BC044799.1 - 5 356 4836807

View Full Product Details