Gene Transcripts

Gene Symbol
STIM1
Entrez Gene ID
6786
Gene Name
stromal interaction molecule 1
Gene Aliases
D11S4896E, GOK, IMD10, STRMK, TAM, TAM1
Chromosome Location
Chr. 11: 3855703 - 4093210 on Build GRCh38
UniGene ID
Hs.501735
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277961.1 NP_001264890.1 2 753
NM_001277962.1 NP_001264891.1 2 753
NM_003156.3 NP_003147.2 2 753
GenBank mRNA AK314928.1 - 2 274
BC021300.2 - 2 745 4899542
HQ317451.1 - 2 752
JX014264.1 - 2 185
U52426.1 - 2 750

View Full Product Details