Gene Transcripts

Gene Symbol
UTP23
Entrez Gene ID
84294
Gene Name
UTP23, small subunit processome component
Gene Aliases
C8orf53
Chromosome Location
Chr. 8: 116766503 - 116774684 on Build GRCh38
UniGene ID
Hs.86970
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032334.2 NP_115710.2 1, 2 283
GenBank mRNA AK055279.1 - 1, 2 232
AK315770.1 - 1, 2 227
BC005955.1 - 1, 2 283 4285502
BC022441.1 - 1, 2 283 4285036
BP231814.1 - 1, 2 283

View Full Product Details