Gene Transcripts
- Gene Symbol
- ZGRF1
- Entrez Gene ID
- 55345
- Gene Name
- zinc finger GRF-type containing 1
- Gene Aliases
- C4orf21
- Chromosome Location
- Chr. 4: 112539333 - 112636995 on Build GRCh38
- UniGene ID
- Hs.380346
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018392.4 |
NP_060862.3
|
12, 13 |
3996 |
|
XM_005263115.3 |
XP_005263172.1
|
12, 13 |
4051 |
|
XM_011532091.2 |
XP_011530393.1
|
11, 12 |
3945 |
|
XM_011532092.2 |
XP_011530394.1
|
11, 12 |
3876 |
|
XM_011532093.2 |
XP_011530395.1
|
11, 12 |
3822 |
|
XM_011532094.2 |
XP_011530396.1
|
10, 11 |
3738 |
|
XM_011532096.2 |
XP_011530398.1
|
12, 13 |
3996 |
|
XM_011532097.2 |
XP_011530399.1
|
5, 6 |
1098 |
|
XM_011532098.2 |
XP_011530400.1
|
3, 4 |
772 |
|
XM_011532099.2 |
XP_011530401.1
|
2, 3 |
731 |
|
XM_017008369.1 |
XP_016863858.1
|
11, 12 |
3991 |
|
XM_017008370.1 |
XP_016863859.1
|
10, 11 |
3771 |
|
XM_017008371.1 |
XP_016863860.1
|
9, 10 |
3651 |
|
GenBank mRNA |
AK126054.1 |
- |
1, 2 |
380 |
|
BX647594.1 |
- |
1, 2 |
479 |
|
CN297341.1 |
- |
1, 2 |
595 |
|
View Full Product Details