Gene Transcripts

Gene Symbol
UBLCP1
Entrez Gene ID
134510
Gene Name
ubiquitin like domain containing CTD phosphatase 1
Gene Aliases
CPUB1
Chromosome Location
Chr. 5: 159263081 - 159286040 on Build GRCh38
UniGene ID
Hs.591733
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145049.3 NP_659486.2 8, 9 1002
GenBank mRNA AK057996.1 - 8, 9 793
AY444562.1 - 8, 9 814
BC013425.2 - 8, 9 814 3894780

View Full Product Details