Gene Transcripts

Gene Symbol
ARMCX4
Entrez Gene ID
100131755
Gene Name
armadillo repeat containing, X-linked 4
Gene Aliases
CXorf35, GASP4
Chromosome Location
Chr. X: 101418263 - 101535988 on Build GRCh38
UniGene ID
Hs.729062
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028407.2 10 2404
NR_045861.1 7 2295
NR_045862.1 6 1429
NR_045863.1 6 1423
NR_045864.1 7 2337
GenBank mRNA AK096955.1 - 8 1785
AK125041.1 - 6 1227
AK126987.1 - 7 2158
BC032236.1 - 7 2074
BC036206.1 - 6 1425
CR933662.1 - 10 2379

View Full Product Details