Gene Transcripts

Gene Symbol
NAV3
Entrez Gene ID
89795
Gene Name
neuron navigator 3
Gene Aliases
POMFIL1, STEERIN3, unc53H3
Chromosome Location
Chr. 12: 77571862 - 78213012 on Build GRCh38
UniGene ID
Hs.655301
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024383.1 NP_001019554.1 12 2722
NM_014903.5 NP_055718.4 12 2722
XM_005269215.3 XP_005269272.1 12 6723
XM_011538940.2 XP_011537242.1 12 6724
XM_011538941.2 XP_011537243.1 12 6723
XM_011538944.2 XP_011537246.1 12 6721
XM_017020164.1 XP_016875653.1 12 6725
XM_017020165.1 XP_016875654.1 12 6725
XM_017020166.1 XP_016875655.1 12 2711
XM_017020167.1 XP_016875656.1 11 2642
XM_017020168.1 XP_016875657.1 12 6724
XM_017020169.1 XP_016875658.1 12 6723
XM_017020170.1 XP_016875659.1 12 6720
XM_017020171.1 XP_016875660.1 4 6018
XM_017020172.1 XP_016875661.1 4 1656
XM_017020173.1 XP_016875662.1 4 6016
XM_017020174.1 XP_016875663.1 4 6016
XM_017020175.1 XP_016875664.1 4 1895
GenBank mRNA AF397731.1 - 11 2306
AJ488201.1 - 12 2594

View Full Product Details