Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003334.3 | NP_003325.2 | 5 | 591 | |
NM_153280.2 | NP_695012.1 | 5 | 519 | ||
XM_005272649.1 | XP_005272706.1 | 6 | 969 | ||
XM_011543954.2 | XP_011542256.1 | 6 | 611 | ||
XM_017029777.1 | XP_016885266.1 | 5 | 1686 | ||
XM_017029778.1 | XP_016885267.1 | 6 | 2860 | ||
XM_017029779.1 | XP_016885268.1 | 5 | 3368 | ||
XM_017029780.1 | XP_016885269.1 | 5 | 476 | ||
XM_017029781.1 | XP_016885270.1 | 5 | 524 | ||
GenBank mRNA | BC013041.2 | - | 5 | 458 | |
BC020261.1 | - | 2 | 153 | ||
BG703038.1 | - | 5 | 537 | ||
BI547207.1 | - | 5 | 597 | ||
M58028.1 | - | 5 | 497 | ||
X56976.1 | - | 5 | 400 |