Gene Transcripts
- Gene Symbol
- ACSS1
- Entrez Gene ID
- 84532
- Gene Name
- acyl-CoA synthetase short-chain family member 1
- Gene Aliases
- ACAS2L, ACECS1, AceCS2L
- Chromosome Location
- Chr. 20: 25006230 - 25058182 on Build GRCh38
- UniGene ID
- Hs.529353
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252675.1 |
NP_001239604.1
|
5, 6 |
1048 |
|
NM_001252676.1 |
NP_001239605.1
|
4, 5 |
875 |
|
NM_001252677.1 |
NP_001239606.1
|
5, 6 |
1048 |
|
NM_032501.3 |
NP_115890.2
|
5, 6 |
1048 |
|
XM_011529388.1 |
XP_011527690.1
|
4, 5 |
833 |
|
GenBank mRNA |
AK092295.1 |
- |
3, 4 |
457 |
|
AK125058.1 |
- |
3, 4 |
2251 |
|
AK127566.1 |
- |
5, 6 |
994 |
|
AK296306.1 |
- |
4, 5 |
872 |
|
AK314440.1 |
- |
5, 6 |
971 |
|
BC039261.1 |
- |
5, 6 |
1007 |
5298849 |
BC044588.1 |
- |
5, 6 |
973 |
5260496 |
BC055008.1 |
- |
5, 6 |
973 |
4838976 |
BC114612.1 |
- |
5, 6 |
839 |
|
View Full Product Details