Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005268208.1 | XP_005268265.1 | 5 | 706 | |
XM_005268209.3 | XP_005268266.1 | 5 | 510 | ||
XM_017021765.1 | XP_016877254.1 | 5 | 509 | ||
GenBank mRNA | AB082526.1 | - | 5 | 678 | |
AK128693.1 | - | 6 | 642 | ||
AY048580.1 | - | 5 | 572 | ||
AY080896.1 | - | 5 | 584 | ||
BC093881.1 | - | 5 | 653 | 7939726 | |
BC112101.1 | - | 5 | 653 | 8327569 |